EMAIL THIS PAGE TO A FRIEND
05033 Sigma-Aldrich

Aflatoxin B2

from Aspergillus flavus

  • CAS Number 7220-81-7

  • Empirical Formula (Hill Notation) C17H14O6

  • Molecular Weight 314.29

  •  Beilstein Registry Number 1355115

  •  EC Number 230-618-8

  •  MDL number MFCD00078140

  •  PubChem Substance ID 329748012

Purchase

Properties

biological source   from Aspergillus flavus
InChI Key   WWSYXEZEXMQWHT-WNWIJWBNSA-N
storage temp.   2-8°C

Description

Warning

Possibly carcinogenic.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGI
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?