EMAIL THIS PAGE TO A FRIEND
05036 Sigma-Aldrich

Aflatoxin G2

from Aspergillus flavus, ≥98.0% (HPLC/TLC)

  • CAS Number 7241-98-7

  • Empirical Formula (Hill Notation) C17H14O7

  • Molecular Weight 330.29

  •  Beilstein Registry Number 1692017

  •  EC Number 230-643-4

  •  MDL number MFCD00078141

Purchase

Properties

biological source   from Aspergillus flavus
InChI Key   WPCVRWVBBXIRMA-WNWIJWBNSA-N
assay   ≥98.0% (HPLC/TLC)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Warning

Possibly carcinogenic.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3462 6.1/PG 1
WGK Germany 
3
RTECS 
LV1700000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

A0263 Aflatoxin G2

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?