EMAIL THIS PAGE TO A FRIEND
05113 Sigma-Aldrich

Activated charcoal

for GC

Synonym: Charcoal activated

Purchase

Properties

Related Categories Activated Charcoal, Adsorbents, Filter Aids and Drying Agents, Essential Chemicals, Research Essentials
grade   for GC
vapor pressure   <0.1 mmHg ( 20 °C)
InChI Key   OKTJSMMVPCPJKN-UHFFFAOYSA-N
autoignition temp.   842 °F
resistivity   1375 μΩ-cm, 20°C (graphite)
application(s)   gas chromatography (GC): suitable
particle size   0.5-1 mm
mp   3550 °C(lit.)
bulk density   ~0.45 g/cm3

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?