• USA Home
  • 05485 - Albumin methylated from bovine serum

EMAIL THIS PAGE TO A FRIEND
05485 Sigma-Aldrich

Albumin methylated from bovine serum

≥90% protein basis

Purchase

Properties

assay   ≥90% protein basis
impurities   ≤10% water
solubility   H2O: soluble50 mg/mL, hazy, light yellow
storage temp.   2-8°C

Description

Other Notes

Sales restrictions may apply

Preparation Note

from 05482 by esterification with methanol.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?