• USA Home
  • 05519 - Fmoc-4-phenyl-piperidine-4-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
05519 Sigma-Aldrich

Fmoc-4-phenyl-piperidine-4-carboxylic acid

≥97.0%

Synonym: Fmoc-4-phenyl-isonipecotic acid

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Other Unnatural Amino Acids & Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   JWEBOBGECOQGTL-UHFFFAOYSA-N
assay   ≥97.0%
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?