• USA Home
  • 05645 - Alcohol Dehydrogenase from equine liver

EMAIL THIS PAGE TO A FRIEND
05645 Sigma-Aldrich

Alcohol Dehydrogenase from equine liver

suspension in 20 mM potassium phosphate buffer, pH 7; ethanol 10%, ampule, 10% ethanol basis, ~30 U/mL

Synonym: ADH, Alcohol Dehydrogenase from horse liver, Alcohol:NAD+ oxidoreductase, HLADH

Purchase

Properties

Related Categories 1.1.x.x Acting on hydroxyl groups, 1.x.x.x Oxidoreductases, Alcohol Metabolism, Application Index, Biochemicals and Reagents,
assay   10% ethanol basis
quality   ampule
  suspension in 20 mM potassium phosphate buffer, pH 7; ethanol 10%
mol wt   Mr ~80000
concentration   ~30 U/mL
pH   7
foreign activity   lactate dehydrogenase ≤0.1%
storage temp.   2-8°C

Description

Other Notes

For the enzymatic determination of ethanol; Dehydrogenases for the biotransformations: Review; Alcohol dehydrogenase-coupled spectrophotometric assay of plasmalogenase

Sales restrictions may apply

Unit Definition

1 U corresponds to the amount of enzyme, which oxidizes 1 μmol ethanol per minute at pH 8.8 and 25°C

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?