EMAIL THIS PAGE TO A FRIEND
05753 Sigma-Aldrich

L-allo-Threonine

puriss., ≥99.0% (TLC)

Synonym: (2S,3S)-2-Amino-3-hydroxybutyric acid

  • CAS Number 28954-12-3

  • Linear Formula CH3CH(OH)CH(NH2)CO2H

  • Molecular Weight 119.12

  •  Beilstein Registry Number 1721645

  •  EC Number 249-327-2

  •  MDL number MFCD00064268

  •  PubChem Substance ID 329748163

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Threonine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   AYFVYJQAPQTCCC-HRFVKAFMSA-N
grade   puriss.
assay   ≥99.0% (TLC)
optical activity   [α]20/D +33±2°, c = 1% in 1 M HCl
mp   272 °C (dec.)(lit.)
  ~275 °C (dec.)

Description

Other Notes

Sales restrictions may apply

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?