EMAIL THIS PAGE TO A FRIEND
05758 Sigma-Aldrich

Alloxan monohydrate

≥98.0% (TLC)

Synonym: 2,4,5,6(1H,3H)-Pyrimidinetetrone, 2,4,5,6-Tetraoxypyrimidine, 5,6-Dioxyuracil

  • CAS Number 2244-11-3

  • Empirical Formula (Hill Notation) C4H2N2O4 · H2O

  • Molecular Weight 160.08

  •  Beilstein Registry Number 4573597

  •  MDL number MFCD00149399

  •  PubChem Substance ID 329748165

Purchase

Properties

Related Categories Building Blocks, C4 to C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
InChI Key   DSXMTJRUNLATRP-UHFFFAOYSA-N
assay   ≥98.0% (TLC)
impurities   ~12% water
mp   255 °C (dec.)(lit.)
solubility   H2O: 50 mg/mL clear
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
UW0492000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A7413 Alloxan monohydrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?