EMAIL THIS PAGE TO A FRIEND
05760 Sigma-Aldrich

Alloxan tetrahydrate

purum p.a., for the detection of Ag, Cd, Co, Fe, HCN, Hg, Mg, Mn, Ni, Zn, ≥98.0% (TLC)

Synonym: 2,4,5,6(1H,3H)-Pyrimidinetetrone, 2,4,5,6-Tetraoxypyrimidine, 5,6-Dioxyuracil

  • CAS Number 6010-91-9

  • Empirical Formula (Hill Notation) C4H2N2O4 · 4H2O

  • Molecular Weight 214.13

  •  Beilstein Registry Number 132451

  •  MDL number MFCD00149400

  •  PubChem Substance ID 329748166

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Decomposition Reagents, Indicators,
InChI Key   JJROYYCRHFNFNC-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥98.0% (TLC)
quality   for the detection of Ag, Cd, Co, Fe, HCN, Hg, Mg, Mn, Ni, Zn
impurities   30-35% water
mp   ~245 °C (dec.)
storage temp.   2-8°C

Description

Other Notes

For the experimental induction of diabetes; Inhibitor of mitochondrial aconitase

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A6316 Alloxan tetrahydrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?