EMAIL THIS PAGE TO A FRIEND
05810 Sigma-Aldrich

Allylamine

purum, ≥98.0% (GC)

Synonym: 2-Propen-1-ylamine, 3-Amino-1-propene

  • CAS Number 107-11-9

  • Linear Formula CH2=CHCH2NH2

  • Molecular Weight 57.09

  •  Beilstein Registry Number 635703

  •  EC Number 203-463-9

  •  MDL number MFCD00008199

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
vapor density   2 (vs air)
vapor pressure   4.09 psi ( 20 °C)
InChI Key   VVJKKWFAADXIJK-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
expl. lim.   22 %
refractive index   n20/D 1.420(lit.)
  n20/D 1.420
bp   53 °C(lit.)
mp   −88 °C(lit.)
density   0.761 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2334 3(6.1) / PGI
WGK Germany 
2
RTECS 
BA5425000
Flash Point(F) 
-20.2 °F
Flash Point(C) 
-29 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

145831 Allylamine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?