EMAIL THIS PAGE TO A FRIEND
05820 Sigma-Aldrich

4-Allylanisole

purum, ≥97.0% (GC)

Synonym: Chavicol methyl ether, Estragole

  • CAS Number 140-67-0

  • Linear Formula H2C=CHCH2C6H4OCH3

  • Molecular Weight 148.20

  •  Beilstein Registry Number 1099454

  •  EC Number 205-427-8

  •  MDL number MFCD00008653

  •  PubChem Substance ID 329748174

Purchase

Properties

Related Categories Acyclic, Alkenes, Artemisia vulgaris, Building Blocks, Cell Biology,
InChI Key   ZFMSMUAANRJZFM-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.521(lit.)
  n20/D 1.521
bp   215-216 °C(lit.)
density   0.965 g/mL at 25 °C(lit.)

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
BZ8225000
Flash Point(F) 
177.8 °F
Flash Point(C) 
81 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A29208 4-Allylanisole, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?