EMAIL THIS PAGE TO A FRIEND
05870 Sigma-Aldrich

Allyl bromide

purum, ≥98.0% (GC)

Synonym: 3-Bromo-1-propene

  • CAS Number 106-95-6

  • Linear Formula CH2=CHCH2Br

  • Molecular Weight 120.98

  •  Beilstein Registry Number 605308

  •  EC Number 203-446-6

  •  MDL number MFCD00000244

  •  PubChem Substance ID 24845882

Purchase

Properties

Related Categories Chemical Synthesis, Other Protecting and Derivatizing Reagents, Protecting and Derivatizing Reagents, Protection and Derivatization, Synthetic Reagents More...
vapor density   4.2 (vs air)
InChI Key   BHELZAPQIKSEDF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   554 °F
contains   silverwool as stabilizer
expl. lim.   7.3 %
refractive index   n20/D 1.469(lit.)
  n20/D 1.469
bp   70-71 °C(lit.)
mp   −119 °C(lit.)
density   1.398 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN1099 - class 3 - PG 1 - Allyl bromide
WGK Germany 
2
RTECS 
UC7090000
Flash Point(F) 
30.2 °F
Flash Point(C) 
-1 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

337528 Allyl bromide, reagent grade, 97%, contains ≤1000 ppm propylene oxide as stabilizer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?