EMAIL THIS PAGE TO A FRIEND
06019 Sigma-Aldrich

Allyl isothiocyanate

purum, ≥98% (GC)

Synonym: AITC, Oil of mustard

  • CAS Number 57-06-7

  • Linear Formula CH2=CHCH2NCS

  • Molecular Weight 99.15

  •  Beilstein Registry Number 773748

  •  MDL number MFCD00004822

  •  PubChem Substance ID 329748191

Purchase

Properties

Related Categories Armoracia rusticana (Horseradish), Building Blocks, Cell Biology, Chemical Synthesis, Nutrition Research,
InChI Key   ZOJBYZNEUISWFT-UHFFFAOYSA-N
grade   purum
assay   ≥98% (GC)
contains   0.1% BHT as stabilizer
refractive index   n20/D 1.529(lit.)
  n20/D 1.530
bp   150 °C(lit.)
mp   −80 °C(lit.)
density   1.013 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Biochem/physiol Actions

Naturally occurring constituent of cruciferous vegetables that has anti-tumor and anti-oxidant properties. Induces the activity of phase II detoxification enzymes in the urinary bladder and, thus, may be chemoprotective against bladder cancer.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1545 3(6.1) / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

377430 Allyl isothiocyanate, contains BHT as stabilizer, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?