• USA Home
  • 06022 - Diethyl allylmalonate

EMAIL THIS PAGE TO A FRIEND
06022 Sigma-Aldrich

Diethyl allylmalonate

purum, ≥95.0% (GC)

Synonym: Allylmalonic acid diethyl ester

  • CAS Number 2049-80-1

  • Linear Formula CH2=CHCH2CH(COOC2H5)2

  • Molecular Weight 200.23

  •  Beilstein Registry Number 1709466

  •  EC Number 218-072-9

  •  MDL number MFCD00009155

Purchase

Properties

Related Categories Building Blocks, C10 to C11, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   GDWAYKGILJJNBB-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.431(lit.)
  n20/D 1.431
bp   222-223 °C(lit.)
density   1.015 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
159.8 °F
Flash Point(C) 
71 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?