• USA Home
  • 06046 - N-(Allyloxycarbonyl)ethanolamine

EMAIL THIS PAGE TO A FRIEND
06046 Sigma-Aldrich

N-(Allyloxycarbonyl)ethanolamine

≥98.0% (GC)

Synonym: Allyl N-(2-hydroxyethyl)carbamate

  • CAS Number 66471-00-9

  • Linear Formula CH2=CHCH2OCONHCH2CH2OH

  • Molecular Weight 145.16

  •  Beilstein Registry Number 1766386

  •  MDL number MFCD01862885

  •  PubChem Substance ID 329748198

Purchase

Properties

Related Categories Amino Alcohols, Bifunctional Linkers, Building Blocks, Chemical Biology, Chemical Synthesis,
InChI Key   GIYCJRYESIQCEW-UHFFFAOYSA-N
assay   ≥98.0% (GC)
refractive index   n20/D 1.471
density   1.125 g/mL at 20 °C(lit.)

Description

Other Notes

Building block for introducing an Alloc-protected 2-aminoethoxy group

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?