• USA Home
  • 06048 - Allyl methyl carbonate

EMAIL THIS PAGE TO A FRIEND
06048 Sigma-Aldrich

Allyl methyl carbonate

purum, ≥98.0% (GC)

  • CAS Number 35466-83-2

  • Linear Formula CH2=CHCH2OCOOCH3

  • Molecular Weight 116.12

  •  Beilstein Registry Number 1746960

  •  MDL number MFCD00134545

  •  PubChem Substance ID 329748199

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   YHLVIDQQTOMBGN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.406(lit.)
  n20/D 1.406
bp   59-60 °C/35 mmHg(lit.)
density   1.022 g/mL at 25 °C(lit.)

Description

Other Notes

Reagent for allylation reactions; Reagent for dehydrogenations

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
91.4 °F
Flash Point(C) 
33 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?