• USA Home
  • 06049 - Allyl(dichloro)methylsilane

EMAIL THIS PAGE TO A FRIEND
06049 Sigma-Aldrich

Allyl(dichloro)methylsilane

purum, ≥95.0% (GC)

Synonym: Allyl-methyl-dichlorosilane

  • CAS Number 1873-92-3

  • Linear Formula H2C=CHCH2Si(Cl)2CH3

  • Molecular Weight 155.10

  •  Beilstein Registry Number 1740877

  •  EC Number 217-498-2

  •  MDL number MFCD00045141

  •  PubChem Substance ID 329748200

Purchase

Properties

InChI Key   YCEQUKAYVABWTE-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.440(lit.)
  n20/D 1.440
bp   119.5 °C(lit.)
density   1.067 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2985 8(3) / PGII
WGK Germany 
1
Flash Point(F) 
68 °F
Flash Point(C) 
20 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

447277 Allyl(dichloro)methylsilane, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?