EMAIL THIS PAGE TO A FRIEND
06055 Sigma-Aldrich

Allyl phenyl ether

puriss., ≥98.5% (GC)

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   POSICDHOUBKJKP-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (GC)
refractive index   n20/D 1.522(lit.)
  n20/D 1.522
bp   192 °C(lit.)
density   0.978 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
DA8575000
Flash Point(F) 
145.4 °F
Flash Point(C) 
63 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A35208 Allyl phenyl ether, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?