EMAIL THIS PAGE TO A FRIEND
06064 Sigma-Aldrich

N-Allylthiourea

purum, ≥98.0% (HPLC)

Synonym: 1-Allyl-2-thiourea, Thiosinamine

  • CAS Number 109-57-9

  • Linear Formula CH2=CHCH2NHCSNH2

  • Molecular Weight 116.18

  •  Beilstein Registry Number 1071470

  •  EC Number 203-683-5

  •  MDL number MFCD00004940

  •  PubChem Substance ID 24845795

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thioureas More...
InChI Key   HTKFORQRBXIQHD-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   70-72 °C(lit.)
  70-73 °C
density   1.11 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
YR8050000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

108804 N-Allylthiourea, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?