EMAIL THIS PAGE TO A FRIEND
06079 Sigma-Aldrich

Bromoacetic acid

analytical standard

  • CAS Number 79-08-3

  • Linear Formula BrCH2COOH

  • Molecular Weight 138.95

  •  Beilstein Registry Number 506167

  •  EC Number 201-175-8

  •  MDL number MFCD00002678

  •  PubChem Substance ID 24845797

Purchase

Properties

Related Categories A-B, A-C, Additional Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
InChI Key   KDPAWGWELVVRCH-UHFFFAOYSA-N
assay   ≥99.0% (GC)
shelf life   limited shelf life, expiry date on the label
bp   208 °C(lit.)
mp   46-49 °C
  47-49 °C(lit.)
Gene Information   human ... PTPN6(5777)

Description

Application

standard for determination of haloacetic acids in water by gas chromatography according to DIN 38407-25

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3425 8 / PGII
WGK Germany 
2
RTECS 
AF5950000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?