EMAIL THIS PAGE TO A FRIEND
06080 Sigma-Aldrich

Allyltriphenylsilane

purum, ≥98.0% (HPLC)

  • CAS Number 18752-21-1

  • Linear Formula H2C=CHCH2Si(C6H5)3

  • Molecular Weight 300.47

  •  Beilstein Registry Number 2941926

  •  MDL number MFCD00077644

  •  PubChem Substance ID 329748208

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Other Organosilicon
InChI Key   DXJZZRSMGLGFPW-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   87-90 °C(lit.)
  87-90 °C

Description

Other Notes

Reagent used for the preparation of vinylsilanes (via γ-alkylation of triphenylsilylallyllithium; γ-alkylation is preferred compared to the TMS analogue which gives also α-alkylated products); For a practical and stereoselective synthesis of 1,3-dienes

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?