EMAIL THIS PAGE TO A FRIEND
06101 Sigma-Aldrich

Potassium hydroxide

technical, ≥85%, lumps

Synonym: Caustic potash

Purchase

Properties

vapor pressure   1 mmHg ( 719 °C)
InChI Key   KWYUFKZDYYNOTN-UHFFFAOYSA-M
grade   technical
assay   ≥85%
form   lumps
mp   361 °C(lit.)
anion traces   chloride (Cl-): ≤100 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Fe: ≤20 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1813 8 / PGII
WGK Germany 
1
RTECS 
TT2100000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?