• USA Home
  • 06160 - Aluminum acetylacetonate

EMAIL THIS PAGE TO A FRIEND
06160 Sigma-Aldrich

Aluminum acetylacetonate

purum, crystallized, ≥95.0% (KT)

Synonym: Al(acac)3, Aluminum 2,4-pentanedionate

Purchase

Properties

Related Categories Aluminum, Catalysis and Inorganic Chemistry, Chemical Synthesis More...
InChI Key   KILURZWTCGSYRE-LNTINUHCSA-K
grade   purum
assay   ≥95.0% (KT)
form   solid
quality   crystallized
bp   315 °C(lit.)
mp   190-193 °C(lit.)
  192-195 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3467 6.1 / PGII
WGK Germany 
3
RTECS 
BD2230000
Flash Point(F) 
244.4 °F
Flash Point(C) 
118 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?