EMAIL THIS PAGE TO A FRIEND
06230 Sigma-Aldrich

Aluminum chloride

purum, anhydrous, ≥98.0% (AT)

Purchase

Properties

Related Categories Acids, Acids & Bases, Aluminum, Aluminum Salts, Catalysis and Inorganic Chemistry,
vapor pressure   1 mmHg ( 100 °C)
InChI Key   VSCWAEJMTAWNJL-UHFFFAOYSA-K
grade   purum
assay   ≥98.0% (AT)
form   powder
mp   190 °C(lit.)
cation traces   Fe: ≤3000 mg/kg

Description

Features and Benefits

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Aluminum nitride can be synthesized from the elemnts with AlCl3 added to help facilitate crystallization.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1726 8 / PGII
WGK Germany 
1
RTECS 
BD0525000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?