• USA Home
  • 06437 - Amberlite® 200 Na+ form

EMAIL THIS PAGE TO A FRIEND
06437 Sigma-Aldrich

Amberlite® 200 Na+ form

Na+-form

Purchase

Properties

Related Categories Amberlite, Amberlite Strong Cation Exchangers, Analytical/Chromatography, Cation, Cation Exchange Media,
description   strongly acidic
moisture   46-52%
loss   ~45% loss on drying
matrix   styrene-divinylbenzene (macroreticular)
particle size   18-23 mesh
  600-850 μm
capacity   1.7 meq/mL by wetted bed volume

Description

Application

Strongly acidic cation exchange resin for water conditioning, removal of heavy metals, etc.

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?