EMAIL THIS PAGE TO A FRIEND
06442 Sigma-Aldrich

Amberlite® XAD16N

20-50 mesh

Purchase

Properties

Related Categories Adsorbent Media (Polymeric), Adsorbents (Resins), Amberlite, Amberlite & Amberchrom, Analytical/Chromatography,
loss   ~65% loss on drying
matrix   polystyrene, apolar
particle size   20-50 mesh
pore size   1.82 mL/g pore volume
  90 Å mean pore size
surface area   800 m2/g

Description

General description

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Application

Hydrophobic compounds up to 40,000 MW; antibiotics; pharmaceutical manufacturing; surfactants; bitters; separation of large organic molecules (especially proteins). More efficient than XAD2.

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?