• USA Home
  • 06461 - Amberlite® IRA-900 chloride form

EMAIL THIS PAGE TO A FRIEND
06461 Sigma-Aldrich

Amberlite® IRA-900 chloride form

chloride form, strongly basic, 20-50 mesh

Purchase

Properties

Related Categories Amberlite, Amberlite Strong Anion Exchangers, Analytical/Chromatography, Anion, Anion Exchange Media,
vapor density   <1 (vs air)
autoignition temp.   800 °F
parameter   60 °C max. temp.
  77 °C max. temp.
moisture   58-64%
loss   ~60% loss on drying, 110 °C
matrix   styrene-divinylbenzene (macroreticular)
particle size   20-50 mesh
  650-820 μm
operating pH   0 - 14
capacity   1.0 meq/mL by wetted bed volume

Description

Application

Strongly basic (type I) anion exchange resin for decolorizing intermediate to light solutions, concentrating heparin at neutral pH, removing metal ions, etc.

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?