• USA Home
  • 06477 - Amberlite® IRA404 chloride form

EMAIL THIS PAGE TO A FRIEND
06477 Sigma-Aldrich

Amberlite® IRA404 chloride form

chloride form

Purchase

Properties

Related Categories Amberlite, Amberlite Strong Anion Exchangers, Analytical/Chromatography, Anion, Anion Exchange Media,
moisture   40-47%
loss   ~65% loss on drying, 110 °C
matrix   styrene-divinylbenzene (gel)
particle size   30-45 mesh
capacity   1.4 meq/mL by wetted bed volume

Description

Application

Strongly basic anion exchange resin for the absorption and removal of organic material from water; organic scavenger.

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?