EMAIL THIS PAGE TO A FRIEND
06530 Sigma-Aldrich

Isopentyl formate

≥95.0%

  • CAS Number 110-45-2

  • Linear Formula HCOOCH2CH2CH(CH3)2

  • Molecular Weight 116.16

  •  Beilstein Registry Number 1739893

  •  EC Number 203-769-2

  •  MDL number MFCD00021049

  •  PubChem Substance ID 329748241

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
vapor pressure   10 mmHg ( 17.1 °C)
InChI Key   XKYICAQFSCFURC-UHFFFAOYSA-N
assay   ≥95.0%
expl. lim.   8 %
impurities   pentyl formate
refractive index   n20/D 1.397(lit.)
  n20/D 1.397
bp   123-124 °C(lit.)
density   0.859 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1109 3 / PGIII
WGK Germany 
2
Flash Point(F) 
78.8 °F
Flash Point(C) 
26 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?