• USA Home
  • 06566 - 4-Amidinophenylmethanesulfonyl fluoride hydrochloride

EMAIL THIS PAGE TO A FRIEND
06566 Sigma-Aldrich

4-Amidinophenylmethanesulfonyl fluoride hydrochloride

≥97.0% (AT)

Synonym: 4-Amidinobenzylsulfonyl fluoride hydrochloride, p-APMSF

  • CAS Number 74938-88-8

  • Empirical Formula (Hill Notation) C8H9FN2O2S · HCl

  • Molecular Weight 252.69

  •  Beilstein Registry Number 9083148

  •  MDL number MFCD00078902

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Organic Covalent Modifiers More...
InChI Key   KHLLRHIUKOJXLL-UHFFFAOYSA-N
assay   ≥97.0% (AT)
mp   ~205 °C (dec.)
storage temp.   −20°C

Description

Other Notes

An irreversible inhibitor of serine proteases

Same product — same quality — now Sigma-Aldrich brand

Application

APMSF was used to study the glucagon-receptor interaction; inhibition of cAMP formation led to the conclusion that there was a previously unexpected serine residue in the active site which may operate on the receptor by a mechanism similar to serine proteases.

Biochem/physiol Actions

Irreversible inhibitor of serine proteases with lysine or arginine substrate specificities. Effective concentration 10-100 μM. Frequently used to characterize newly discovered proteases.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?