• USA Home
  • 06640 - 4′-Aminoacetophenone

EMAIL THIS PAGE TO A FRIEND
06640 Sigma-Aldrich

4′-Aminoacetophenone

for spectrophotometric det. of Ce, Pd, ≥98.0% (NT)

Synonym: 4-Acetylaniline

  • CAS Number 99-92-3

  • Linear Formula H2NC6H4COCH3

  • Molecular Weight 135.16

  •  Beilstein Registry Number 471493

  •  EC Number 202-801-2

  •  MDL number MFCD00007896

  •  PubChem Substance ID 329748254

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Spectroscopy, UV/Vis Reagents, UV/Visible (UV/VIS) Spectroscopy More...
InChI Key   GPRYKVSEZCQIHD-UHFFFAOYSA-N
assay   ≥98.0% (NT)
quality   for spectrophotometric det. of Ce, Pd
bp   293 °C(lit.)
mp   103-107 °C(lit.)
  104-106 °C

Description

Other Notes

Reagent for the photometric determination of Ce

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AM5500000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?