EMAIL THIS PAGE TO A FRIEND
06760 Sigma-Aldrich

1-Aminoanthracene

technical, ≥90% (GC/NT)

Synonym: 1-Anthramine

  • CAS Number 610-49-1

  • Empirical Formula (Hill Notation) C14H11N

  • Molecular Weight 193.24

  •  Beilstein Registry Number 2209406

  •  EC Number 210-225-8

  •  MDL number MFCD00003579

  •  PubChem Substance ID 329748275

Purchase

Properties

InChI Key   YUENFNPLGJCNRB-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC/NT)
mp   114-118 °C(lit.)
  ~115 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
CA9100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?