EMAIL THIS PAGE TO A FRIEND
06765 Sigma-Aldrich

2-Aminoanthracene

technical, ≥90% (HPLC)

Synonym: 2-Anthramine

  • CAS Number 613-13-8

  • Empirical Formula (Hill Notation) C14H11N

  • Molecular Weight 193.24

  •  Beilstein Registry Number 2209414

  •  EC Number 210-330-9

  •  MDL number MFCD00003582

  •  PubChem Substance ID 24845968

Purchase

Properties

InChI Key   YCSBALJAGZKWFF-UHFFFAOYSA-N
grade   technical
assay   ≥90% (HPLC)
mp   235-240 °C
  238-241 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
CA9275000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?