EMAIL THIS PAGE TO A FRIEND
06790 Sigma-Aldrich

2-Aminoanthraquinone

technical, 85-90% (HPLC)

  • CAS Number 117-79-3

  • Empirical Formula (Hill Notation) C14H9NO2

  • Molecular Weight 223.23

  •  Beilstein Registry Number 745903

  •  EC Number 204-208-4

  •  MDL number MFCD00001233

  •  PubChem Substance ID 329748281

Purchase

Properties

InChI Key   XOGPDSATLSAZEK-UHFFFAOYSA-N
grade   technical
assay   85-90% (HPLC)
impurities   3-5% 2-chloroanthraquinone
mp   292-295 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
CB5120000
Flash Point(F) 
541 °F
Flash Point(C) 
283 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of a 32-Component Anilines Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

165549 2-Aminoanthraquinone, technical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?