EMAIL THIS PAGE TO A FRIEND
06940 Sigma-Aldrich

4-Aminobenzoic acid

≥98.0% (HPLC/NT)

Synonym: PABA, Vitamin Bx, Vitamin H1

  • CAS Number 150-13-0

  • Linear Formula H2NC6H4CO2H

  • Molecular Weight 137.14

  •  Beilstein Registry Number 471605

  •  EC Number 205-753-0

  •  MDL number MFCD00007894

  •  PubChem Substance ID 24845985

  •  eCl@ss 32160406

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   ALYNCZNDIQEVRV-UHFFFAOYSA-N
assay   ≥98.0% (HPLC/NT)
mp   186-189 °C
  187-189 °C(lit.)
density   1.374 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?