EMAIL THIS PAGE TO A FRIEND
06950 Sigma-Aldrich

Benzocaine

purum, ≥99.0% (HPLC)

Synonym: 4-Aminobenzoic acid ethyl ester, Ethyl 4-aminobenzoate

  • CAS Number 94-09-7

  • Empirical Formula (Hill Notation) C9H11NO2

  • Molecular Weight 165.19

  •  Beilstein Registry Number 638434

  •  EC Number 202-303-5

  •  MDL number MFCD00007892

  •  PubChem Substance ID 329748310

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   BLFLLBZGZJTVJG-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (HPLC)
loss   ≤0.5% loss on drying, 20 °C (HV)
mp   88-90 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DG2450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?