• USA Home
  • 06993 - 3-(1-Pyrazolyl)-L-alanine

EMAIL THIS PAGE TO A FRIEND
06993 Sigma-Aldrich

3-(1-Pyrazolyl)-L-alanine

produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)

Synonym: (S)-2-Amino-3-(1-pyrazolyl)propionic acid

  • CAS Number 2734-48-7

  • Empirical Formula (Hill Notation) C6H9N3O2

  • Molecular Weight 155.15

  •  Beilstein Registry Number 83142

  •  MDL number MFCD03791063

  •  PubChem Substance ID 329748320

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
grade   produced by Wacker Chemie AG, Burghausen, Germany
InChI Key   PIGOPELHGLPKLL-YFKPBYRVSA-N
assay   ≥95.0% (HPLC)
optical purity   ee: >99% (due to stereospecific enzymatic synthesis)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?