EMAIL THIS PAGE TO A FRIEND
07035 Sigma-Aldrich

1-Aminobenzotriazole

≥98.0% (GC)

Synonym: 1-Benzotriazolamine, ABT

  • CAS Number 1614-12-6

  • Empirical Formula (Hill Notation) C6H6N4

  • Molecular Weight 134.14

  •  Beilstein Registry Number 607843

  •  MDL number MFCD00132902

  •  PubChem Substance ID 24845815

Purchase

Properties

Related Categories Benzotriazoles, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
InChI Key   JCXKHYLLVKZPKE-UHFFFAOYSA-N
assay   ≥98.0% (GC)
mp   81-84 °C(lit.)
  81-84 °C
Gene Information   human ... CYP1A2(1544)

Description

Biochem/physiol Actions

Suicide inhibitor of cytochrome P450 and chloroperoxidase.

Other Notes

Starting material for generating benzyne with Pb(OAc)4

Same product — same quality — now Sigma-Aldrich brand

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?