• USA Home
  • 07060 - 3-(Trifluoromethyl)aniline

EMAIL THIS PAGE TO A FRIEND
07060 Sigma-Aldrich

3-(Trifluoromethyl)aniline

technical, ≥90% (GC)

Synonym: α,α,α-Trifluoro-m-toluidine, 3-Aminobenzotrifluoride

  • CAS Number 98-16-8

  • Linear Formula CF3C6H4NH2

  • Molecular Weight 161.12

  •  Beilstein Registry Number 387672

  •  EC Number 202-643-4

  •  MDL number MFCD00007797

  •  PubChem Substance ID 24846003

Purchase

Properties

vapor pressure   0.3 mmHg ( 20 °C)
InChI Key   VIUDTWATMPPKEL-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
refractive index   n20/D 1.480(lit.)
  n20/D 1.480
bp   187 °C(lit.)
mp   5-6 °C(lit.)
density   1.29 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2948 6.1 / PGII
WGK Germany 
2
RTECS 
XU9180000
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?