07102 Sigma-Aldrich

Hydrochloric acid

meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%

DOWNLOAD MSDS (PDF)
  • CAS Number 7647-01-0

  • Empirical Formula (Hill Notation) HCl

  • Molecular Weight 36.46

  •  Beilstein Registry Number 1098214

  •  EC Number 231-595-7

  •  MDL number MFCD00011324

  •  PubChem Substance ID 24846013

Properties

Related Categories Chemical Synthesis, Inorganic Acids, Synthetic Reagents More...
vapor density   1.3 (vs air)
vapor pressure   3.23 psi ( 21.1 °C)
  7.93 psi ( 37.7 °C)
assay   36.5-38%
quality   meets analytical specification of Ph. Eur., BP, NF
impurities   free bromine or chlorine, complies
  residual solvents, complies
  ≤0.0001% free chlorine (Cl)
  ≤0.0001% heavy metals (as Pb)
  ≤0.005% non-volatile matter
ign. residue   ≤0.002% (as SO4)
bp   >100 °C(lit.)
density   1.2 g/mL at 25 °C(lit.)
anion traces   bromide (Br-): in accordance
  iodide (I-): in accordance
  sulfate (SO42-): ≤2 mg/kg
  sulfite (SO32-): ≤5 mg/kg
cation traces   As: ≤0.5 mg/kg
  Fe: ≤1 mg/kg
suitability   complies for appearance of solution

Description

Packaging

In PE-bottles; also available in Glass-bottles (Material no. ending with *-GL), PE canister (Material no. ending with *-H);
1 lt only available in glass btl.

Price and Availability

Customers Also Viewed

Hydrochloric acid

36.5-38.0%, BioReagent, for molecular biology

Hydrochloric acid

ACS reagent, 37%

Hydrochloric acid

ACS reagent, 37%

Hydrochloric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10

Sulfuric acid

puriss., meets analytical specification of Ph. Eur., BP, 95-97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?