• USA Home
  • 07179 - (S)-(+)-2-Amino-1-butanol

EMAIL THIS PAGE TO A FRIEND
07179 Sigma-Aldrich

(S)-(+)-2-Amino-1-butanol

technical, ≥97.0% (sum of enantiomers, GC)

Purchase

Properties

InChI Key   JCBPETKZIGVZRE-BYPYZUCNSA-N
grade   technical
assay   ≥97.0% (sum of enantiomers, GC)
optical activity   [α]20/D +11.5±1°, c = 2% in ethanol
refractive index   n20/D 1.4521(lit.)
  n20/D 1.453
bp   172-174 °C(lit.)
density   0.944 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2735 8/PG 3
WGK Germany 
3
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?