• USA Home
  • 07193 - Tellurium Standard for ICP
EMAIL THIS PAGE TO A FRIEND
07193 Sigma-Aldrich
Tellurium Standard for ICP
TraceCERT®, 1000 mg/L Te in nitric acid
Purchase
Description
Analysis Note
1000 mg/L Te in 2% nitric acid (contains HF traces), prepared with high purity Te metal, HNO3 and water
General description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34. This CRM is traceable to SI unit kg and measured against NIST SRM.
Certified content incl. uncertainty and expiry date are given on label. Information about trace impurities are stated in the enclosed certificate (available as download: http://www.sigma-aldrich.com).
Legal Information
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
78358 Tellurium Standard for ICP, TraceCERT®, 1000 mg/L Te in nitric acid and hydrofluoric acid
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?