• USA Home
  • 07253 - (±)-α-Amino-γ-butyrolactone hydrobromide

EMAIL THIS PAGE TO A FRIEND
07253 Sigma-Aldrich

(±)-α-Amino-γ-butyrolactone hydrobromide

purum, ≥97.0% (AT)

Synonym: DL-Homoserine lactone hydrobromide

  • CAS Number 6305-38-0

  • Empirical Formula (Hill Notation) C4H7NO2 · HBr

  • Molecular Weight 182.02

  •  Beilstein Registry Number 4530084

  •  EC Number 228-614-6

  •  MDL number MFCD00012723

  •  PubChem Substance ID 329748385

Purchase

Properties

Related Categories Amino Acids & Derivatives, Amino Lactones, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   MKLNTBLOABOJFZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   220-225 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A44509 α-Amino-γ-butyrolactone hydrobromide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?