• USA Home
  • 07600 - 3-Aminocrotononitrile

EMAIL THIS PAGE TO A FRIEND
07600 Sigma-Aldrich

3-Aminocrotononitrile

purum, ≥95.0% (T)

Synonym: 3-Amino-2-butenenitrile, 3-Iminobutyronitrile, Diacetonitrile

  • CAS Number 1118-61-2

  • Linear Formula H2N(CH3)=CHCN

  • Molecular Weight 82.10

  •  Beilstein Registry Number 1719815

  •  EC Number 214-266-2

  •  MDL number MFCD00008071

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   DELJOESCKJGFML-DUXPYHPUSA-N
grade   purum
assay   ≥95.0% (T)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3439 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

117641 3-Aminocrotononitrile, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?