• USA Home
  • 07680 - 2-Amino-3,5-dichloropyridine

EMAIL THIS PAGE TO A FRIEND
07680 Sigma-Aldrich

2-Amino-3,5-dichloropyridine

purum, ≥97.0% (AT)

  • CAS Number 4214-74-8

  • Empirical Formula (Hill Notation) C5H4Cl2N2

  • Molecular Weight 163.00

  •  Beilstein Registry Number 119376

  •  EC Number 224-143-5

  •  MDL number MFCD00006313

Purchase

Properties

InChI Key   OCWBGKZFOYMCCN-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   81-83 °C(lit.)
  81-83 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

135925 2-Amino-3,5-dichloropyridine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?