• USA Home
  • 07720 - 2-Amino-4,5-dimethoxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
07720 Sigma-Aldrich

2-Amino-4,5-dimethoxybenzoic acid

technical, ≥97.0% (T)

Synonym: 4,5-Dimethoxyanthranilic acid, 6-Aminoveratric acid

  • CAS Number 5653-40-7

  • Linear Formula H2NC6H2(OCH3)2CO2H

  • Molecular Weight 197.19

  •  Beilstein Registry Number 782814

  •  EC Number 227-095-3

  •  MDL number MFCD00011671

  •  PubChem Substance ID 329748509

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   HJVAVGOPTDJYOJ-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (T)
mp   169-173 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?