• USA Home
  • 07760 - N,N-Dimethyl-p-phenylenediamine

EMAIL THIS PAGE TO A FRIEND
07760 Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine

technical, ≥95.0% (GC)

Synonym: 4-(Dimethylamino)aniline, 4-Amino-N,N-dimethylaniline, N,N-Dimethyl-1,4-phenylenediamine, DMPPDA

  • CAS Number 99-98-9

  • Linear Formula (CH3)2NC6H4NH2

  • Molecular Weight 136.19

  •  Beilstein Registry Number 508105

  •  EC Number 202-807-5

  •  MDL number MFCD00007860

  •  PubChem Substance ID 329748515

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   BZORFPDSXLZWJF-UHFFFAOYSA-N
grade   technical
assay   ≥95.0% (GC)
form   crystalline
color   black
bp   262 °C(lit.)
mp   34-36 °C(lit.)
  34-38 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
ST0874000
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

control valve

Product #

Description

Add to Cart

193992 N,N-Dimethyl-p-phenylenediamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?