• USA Home
  • 07780 - N,N-Dimethyl-p-phenylenediamine sulfate salt

EMAIL THIS PAGE TO A FRIEND
07780 Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine sulfate salt

purum, ≥98.0% (T)

Synonym: 4-(Dimethylamino)aniline sulfate salt, 4-Amino-N,N-dimethylaniline sulfate salt, DMPPDA

  • CAS Number 536-47-0

  • Linear Formula (CH3)2NC6H4NH2·H2SO4

  • Molecular Weight 234.27

  •  Beilstein Registry Number 4612278

  •  EC Number 208-636-2

  •  MDL number MFCD00012992

  •  PubChem Substance ID 329748519

Purchase

Properties

Related Categories Amine Salts, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   GLUKPDKNLKRLHX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
bp   495 °C(lit.)
mp   200-205 °C (dec.)(lit.)

Description

Other Notes

This product has been replaced by 186384-ALDRICH | N,N-Dimethyl-p-phenylenediamine sulfate salt 98%

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

186384 N,N-Dimethyl-p-phenylenediamine sulfate salt, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?