EMAIL THIS PAGE TO A FRIEND
07940 Sigma-Aldrich

Benzhydrylamine

purum, ≥97.0% (GC)

Synonym: α-Aminodiphenylmethane

  • CAS Number 91-00-9

  • Linear Formula (C6H5)2CHNH2

  • Molecular Weight 183.25

  •  Beilstein Registry Number 776434

  •  EC Number 202-032-2

  •  MDL number MFCD00008059

  •  PubChem Substance ID 24846125

Purchase

Properties

InChI Key   MGHPNCMVUAKAIE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.595(lit.)
  n20/D 1.596
bp   295 °C(lit.)
mp   12 °C(lit.)
density   1.063 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DA4407300
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

A53605 Benzhydrylamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?