EMAIL THIS PAGE TO A FRIEND
07942 Sigma-Aldrich

2-Benzylaniline

purum, ≥99.0% (NT)

Synonym: 2-Aminodiphenylmethane

  • CAS Number 28059-64-5

  • Linear Formula C6H5CH2C6H4NH2

  • Molecular Weight 183.25

  •  Beilstein Registry Number 2803264

  •  EC Number 248-806-3

  •  MDL number MFCD00007750

Purchase

Properties

InChI Key   DWOBGCPUQNFAFB-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (NT)
bp   172-173 °C/12 mmHg(lit.)
mp   49-54 °C(lit.)
  49-54 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

235350 2-Benzylaniline, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?